Learn More
Cayman Chemical VEGFR2 KInase InhIbItr II 5mg
SDP
Catalog No. 502966102
Encompass_Preferred
A reversible, cell-permeable inhibitor of VEGFR2’s kinase activity (IC50 = 70 nM); less potently inhibits PDGFRβ (IC50 = 920 nM); blocks the growth of human umbilical vein endothelial cells stimulated with either VEGF or PDGF (IC50s = 110 nM and 2 µM, respectively)
Catalog No. 50-296-6102 Supplier Cayman Chemical Supplier No. 175445 MG
Add to Cart
Add to Cart
A reversible, cell-permeable inhibitor of VEGFR2’s kinase activity (IC50 = 70 nM); less potently inhibits PDGFRβ (IC50 = 920 nM); blocks the growth of human umbilical vein endothelial cells stimulated with either VEGF or PDGF (IC50s = 110 nM and 2 µM, respectively)