Learn More
ACROBiosystems ACROBiosystems Human VEGF R2 / KDR Protein, Strep Tag
Catalog No. 501957183
Encompass_Preferred
Recombinant Protein;1MG;Human VEGF R2, Strep Tag (KDR-H5280) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 86.3 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF121, Tag Free (Cat. No. VE1-H4213) at 2 μg/mL (100 μL/well) can bind Human VEGF R2, Strep Tag (Cat. No. KDR-H5280) with a linear range of 0.15-2.5 μg/mL (QC tested).
Catalog No. 50-195-7183 Supplier ACROBiosystems Supplier No. KDRH52801MG
Recombinant Protein;1MG;Human VEGF R2, Strep Tag (KDR-H5280) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823).;This protein carries a twin strep tag at the C-terminus. The protein has a calculated MW of 86.3 kDa. The protein migrates as 100-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF121, Tag Free (Cat. No. VE1-H4213) at 2 μg/mL (100 μL/well) can bind Human VEGF R2, Strep Tag (Cat. No. KDR-H5280) with a linear range of 0.15-2.5 μg/mL (QC tested).