Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque VEGF R2 / KDR Protein;HEK293;100UG;VE2,KDR,CD309,FLK1,VEGFR,VEGFR2,VEGF R
Catalog No. 501658560
Encompass_Preferred
Recombinant Protein;Rhesus macaque VEGF R2 / KDR Protein;100UG;Rhesus macaque VEGF R2, His Tag (VE2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # F7E313).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF165 (Cat.No. VE5-H4210) at 2 g/mL (100 L/well) can bind Rhesus macaque VEGF R2, His Tag (Cat.No. VE2-C52H3) with a linear range of 4-60 ng/mL (QC tested).
Catalog No. 50-165-8560 Supplier ACROBiosystems Supplier No. VE2C52H3100UG
Recombinant Protein;Rhesus macaque VEGF R2 / KDR Protein;100UG;Rhesus macaque VEGF R2, His Tag (VE2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # F7E313).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized ActiveMax Human VEGF165 (Cat.No. VE5-H4210) at 2 g/mL (100 L/well) can bind Rhesus macaque VEGF R2, His Tag (Cat.No. VE2-C52H3) with a linear range of 4-60 ng/mL (QC tested).