Learn More
ACROBiosystems Recombinant Protein;Human VEGF R2 / KDR Protein (HPLC-verified);HEK293;1MG;KDR,KDR,CD309,FLK1,VEGFR,VEGFR2,VEGF R
Catalog No. 501657680
Encompass_Preferred
Recombinant Protein;Human VEGF R2 / KDR Protein (HPLC-verified);1MG;Human VEGF R2, His Tag (KDR-H5227) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Human VEGF R2, His Tag (Cat. No. KDR-H5227) with a linear range of 10-156 ng/mL (QC tested).
Catalog No. 50-165-7680 Supplier ACROBiosystems Supplier No. KDRH52271MG
Recombinant Protein;Human VEGF R2 / KDR Protein (HPLC-verified);1MG;Human VEGF R2, His Tag (KDR-H5227) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 764 (Accession # AAI31823.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 85.2 kDa. The protein migrates as 125-150 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Human VEGF R2, His Tag (Cat. No. KDR-H5227) with a linear range of 10-156 ng/mL (QC tested).