Learn More
Sino Biological Human VEGFR2 / Flk-1 / CD309 / KDR Protein (Domain 1&2&3, Fc Tag), Biotinylated 20ug
Supplier:  Sino Biological 10012H02H1B20
Encompass_Preferred
A DNA sequence encoding the human KDR (NP_002244.1) (Met1-Lys327) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5389