Learn More
ACROBiosystems Recombinant Protein;Mouse VEGF R2 / KDR Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;VE2,KDR,CD309,FLK1,VEGFR,VEGFR2,VEGF R
Catalog No. 501658124
Encompass_Preferred
Recombinant Protein;Mouse VEGF R2 / KDR Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse VEGF R2, mouse IgG2a Fc Tag, low endotoxin (VE2-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 762 (Accession # P35918-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 110.1 kDa. The protein migrates as 120-135 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, mouse IgG2a Fc Tag (Cat. No. VE2-M5258) with a linear range of 1-31 ng/mL (QC tested).;Immobilized Mouse VEGF120, Tag Free (Cat. No. VE0-M4211) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, mouse IgG2a Fc Tag (Cat. No. VE2-M5258) with a linear range of 1-31 ng/mL (Routinely tested).
Catalog No. 50-165-8124 Supplier ACROBiosystems Supplier No. VE2M5258100UG
Recombinant Protein;Mouse VEGF R2 / KDR Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse VEGF R2, mouse IgG2a Fc Tag, low endotoxin (VE2-M5258) is expressed from human 293 cells (HEK293). It contains AA Ala 20 - Glu 762 (Accession # P35918-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 110.1 kDa. The protein migrates as 120-135 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF165, Tag Free (Cat. No. VE5-H4210) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, mouse IgG2a Fc Tag (Cat. No. VE2-M5258) with a linear range of 1-31 ng/mL (QC tested).;Immobilized Mouse VEGF120, Tag Free (Cat. No. VE0-M4211) at 2 μg/mL (100 μL/well) can bind Mouse VEGF R2, mouse IgG2a Fc Tag (Cat. No. VE2-M5258) with a linear range of 1-31 ng/mL (Routinely tested).