Learn More
ACROBiosystems Recombinant Protein;Mouse VEGF R3 / FLT4 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;FL4,Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL,VEGF R3
Supplier:  ACROBiosystems FL4M5251100UG
Encompass_Preferred
Recombinant Protein;Mouse VEGF R3 / FLT4 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (FL4-M5251) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Asp 770 (Accession # P35917-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 111.5 kDa. The protein migrates as 116-125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VEGF-C, His Tag (Cat. No. VEC-H4225) at 5 μg/mL (100 μL/well) can bind Mouse VEGF R3, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. FL4-M5251) with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-165-7974