Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio-Serv™ Veggie-Bites™, Certified
SDP

Catalog No. 14726696
Encompass
Change view
Click to view available options
Quantity:
2 x 5 kg bags/box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-726-696 2 x 5 kg bags/box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-726-696 Supplier Bio-Serv™ Supplier No. F5158
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Used as feed during training reward or foraging treat.

  • Certified (Contaminant Screened)
  • All natural, crunchy vegetable treat
  • Contains mix of dehydrated sliced carrots, puffed peas, and whole kernel corn
  • Less labor intensive and more sanitary than perishable vegetables
  • Nutritionally Assayed

Note: This product in no longer gamma irradiated. It is now Certified (Contaminant Screened). If you require gamma irradiation, please contact customer service for pricing and lead time.

Specifications

Product Type Animal Treats
Calories 3.44 kcal/g
Components Dehydrated Carrots, Puffed Peas, Dried Whole Kernel Corn
Format Dehydrated
For Use With (Application) Feed as a training reward or foraging treat
Quantity 2 x 5 kg bags/box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.