Learn More
Brady™ Vehicle Conspicuity Tape
Catalog No. 19104411 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
DOT-C2 Red
DOT-C2 White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-104-411 DOT-C2 Red
19-104-410 DOT-C2 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-411 Supplier Brady™ Supplier No. 76435
  • $1,509.00
    $915.65 / Each of 1
    Save $593.35
    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
The National Highway Traffic Safety Administration requires all trailers larger than 30 inches wide with a minimum gross weight of 20,000 lbs be equipped on the sides and rear with means for increasing their conspicuity. Our vehicle conspicuity tape is the perfect solution for this rule.
  • DOT-C2
  • Measures 2 in. H x 50 yd L
Specifications
Color DOT-C2 Red
Packaging Quantity 50 yd./Roll
Displayed Text Blank
Height (English) 2 in.
Length (English) 50 yd.
Thickness 0.022 in.
Material Retroreflective Sheeting
Type Marking Tape