Learn More
ATCC Veillonella atypica
Catalog No. 502383827
Encompass
Veillonella atypica strain KON is a whole-genome sequenced bacterial type strain that was isolated from a human mouth.
Catalog No. 50-238-3827 Supplier ATCC Supplier No. 17744
Veillonella atypica strain KON is a whole-genome sequenced bacterial type strain that was isolated from a human mouth.