Learn More
Braintree Scientific Veinlite TV
SDP
Catalog No. 501956095
Encompass
Trans-illuminates veins in rats and mice. Adapter secures tail for visualization of veins and arteries. Comes complete with one mouse and one rat adapter. Click to Watch Video **Sold for research purposes only and not for human use**
Catalog No. 50-195-6095 Supplier Braintree Scientific Supplier No. VLTV
Trans-illuminates veins in rats and mice. Adapter secures tail for visualization of veins and arteries. Comes complete with one mouse and one rat adapter. Click to Watch Video **Sold for research purposes only and not for human use**