Learn More
MSC VELAB Hanging Scale, 100 kg, 200 lb, LCD Digital
Catalog No. 5000527733
Encompass
The VELAB VE-HSS100 is a digital hanging scale designed for accurate weight measurement. It features a robust design with a capacity of 100 kg (200 lb) and an easy-to-read LCD display, making it suitable for various industrial and laboratory applications.
  • Digital hanging scale for precise measurements.
  • Capacity of 100 kg (200 lb).
  • Clear LCD display for easy reading.
  • Hold function retains weight value, aiding in data recording.
  • Graduation of .001 for detailed readings.
Catalog No. 50-005-27733 Supplier MSC Supplier No. 29317096
The VELAB VE-HSS100 is a digital hanging scale designed for accurate weight measurement. It features a robust design with a capacity of 100 kg (200 lb) and an easy-to-read LCD display, making it suitable for various industrial and laboratory applications.
  • Digital hanging scale for precise measurements.
  • Capacity of 100 kg (200 lb).
  • Clear LCD display for easy reading.
  • Hold function retains weight value, aiding in data recording.
  • Graduation of .001 for detailed readings.