Learn More
Medchemexpress LLC Velagliflozin proline | 1539295-26-5 | 99.4% | 510.58 | 1 ML
SDP
Catalog No. 5000346631
Encompass_Preferred
Velagliflozin proline is an oral sodium-glucose cotransporter 2 (SGLT2) inhibitor that exhibits antidiabetic activity. It functions by decreasing renal glucose reabsorption and promoting glycosuria, which consequently leads to a reduction in blood sugar and insulin concentrations.
  • Increases cholesterol, albumin, beta-hydroxybutyrate, nonesterified fatty acids, and urinary glucose excretion.
  • Decreases respiratory exchange ratio.
  • Is well tolerated.
  • Can improve insulin disorders.
  • Prevents laminitis by reducing the high insulin response of dietary non-structural carbohydrates.
Catalog No. 50-003-46631 Supplier Medchemexpress LLC Supplier No. HY109018A10MM/1ML
Velagliflozin proline is an oral sodium-glucose cotransporter 2 (SGLT2) inhibitor that exhibits antidiabetic activity. It functions by decreasing renal glucose reabsorption and promoting glycosuria, which consequently leads to a reduction in blood sugar and insulin concentrations.
  • Increases cholesterol, albumin, beta-hydroxybutyrate, nonesterified fatty acids, and urinary glucose excretion.
  • Decreases respiratory exchange ratio.
  • Is well tolerated.
  • Can improve insulin disorders.
  • Prevents laminitis by reducing the high insulin response of dietary non-structural carbohydrates.