Learn More
Pyramex™ Velar™ Safety Eyewear
Catalog No. 19555707 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Frame Color:
Black
White, Red
Lens Tint:
Blue, Mirror
Clear
Gray
Red, Mirror
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Tint
19-555-707 White, Red Red, Mirror
19-555-670 Black Blue, Mirror
19-555-668 Black Clear
19-555-669 Black Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-555-707 Supplier Pyramex™ Supplier No. SWR10455D
Add to Cart
Add to Cart
This item is not returnable. View return policy
Pyramex™ Velar Safety Eyewear with ANSI Z87.1 safety compliance
  • Built-in rubber nosepiece
  • Co-injected rubber temples
  • Blue Mirror Lens is a gray polycarbonate lens with a double layer of silver, and blue mirror coatings and are commonly used in outdoor applications
  • Lens Technology:
    • Visible Light Transmitted - 13%
    • UVA, UVB Blocked - 99%
    • Blue Light Blocked - 95%
    • IR Blocked - 51%
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance ANSI Z87.1
Lens Tint Red, Mirror
For Use With (Application) Eye Safety
Frame Color White, Red
Temple Style Co-Injected Rubber
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection UV400