Learn More
Enterprise Technology Solutions Velcro Industries Fastener,Hook&Loop,High-Strength,Xtreme Temp.,140F,1",Black
SDP
Catalog No. 5000824181
Encompass_Preferred
High-Strength Fastener lets you complete mounting projects in demanding environments with ease. Use in areas with vibration, dust or more extreme temperature variation. When used indoors, hook-and-loop closure with medium profile delivers two times the strength of General-Use Fasteners. Adhesive works suitably for a wide range of substrates and temperatures (140 degrees Fahrenheit).
Catalog No. 50-008-24181 Supplier Enterprise Technology Solutions Supplier No. VEK31306
High-Strength Fastener lets you complete mounting projects in demanding environments with ease. Use in areas with vibration, dust or more extreme temperature variation. When used indoors, hook-and-loop closure with medium profile delivers two times the strength of General-Use Fasteners. Adhesive works suitably for a wide range of substrates and temperatures (140 degrees Fahrenheit).