Learn More
Medchemexpress LLC Veldoreotide TFA | 2126831-23-8 | 99.0% | 1237.33 | 5 MG
SDP
Catalog No. 5000425029
Encompass_Preferred
Veldoreotide TFA (DG3173) is a somatostatin analogue that binds to and activates somatostatin receptors (SSTR) 2, 4, and 5. It inhibits growth hormone (GH) secretion in adenomas and has potential use as a pain modulating agent.
  • Activates somatostatin receptors 2, 4, and 5
  • Inhibits growth hormone (GH) secretion in adenomas
  • May be used as a pain-modulating agent
  • Stimulates SST2, SST4, and SST5 receptors in HEK293 cells co-expressing these receptors with GIRK2 channels
  • Inhibits SST4-expressing BON-1 cells at 10 μM for 24 hours
Catalog No. 50-004-25029 Supplier Medchemexpress LLC Supplier No. HYP0024A5MG
Veldoreotide TFA (DG3173) is a somatostatin analogue that binds to and activates somatostatin receptors (SSTR) 2, 4, and 5. It inhibits growth hormone (GH) secretion in adenomas and has potential use as a pain modulating agent.
  • Activates somatostatin receptors 2, 4, and 5
  • Inhibits growth hormone (GH) secretion in adenomas
  • May be used as a pain-modulating agent
  • Stimulates SST2, SST4, and SST5 receptors in HEK293 cells co-expressing these receptors with GIRK2 channels
  • Inhibits SST4-expressing BON-1 cells at 10 μM for 24 hours