Learn More
TARGETMOL CHEMICALS INC VELIPARIB 25MG
SDP
Catalog No. 502577087
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Veliparib (ABT-888) (ABT-888) is an orally bioavailable inhibitor of PARP (Kis 5.2/2.9 nM for PARP1/2). It enhances apoptosis and autophagy. purity: 99%
Catalog No. 50-257-7087 Supplier TARGETMOL CHEMICALS INC Supplier No. T259125MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Veliparib (ABT-888) (ABT-888) is an orally bioavailable inhibitor of PARP (Kis 5.2/2.9 nM for PARP1/2). It enhances apoptosis and autophagy. purity: 99%