Learn More
Medchemexpress LLC Veliparib | 912444-00-9 | MFCD16661059 | 244.29 g/mol | C13H16N4O | 200 MG
SDP
Catalog No. 5000212502
Encompass_Preferred
Veliparib (ABT-888) is a small-molecule inhibitor of poly(ADP-ribose) polymerases used in research to probe DNA repair pathways and for chemosensitization studies. It is supplied as a solid for analytical and preclinical research and has documented storage recommendations for powder and solutions.
  • Potent PARP1 and PARP2 inhibition (Ki 5.2 nM and 2.9 nM).
  • Molecular formula C13H16N4O and molecular weight 244.29 g/mol.
  • Identified by CAS number 912444-00-9.
  • Supplied as a powder with recommended storage: -20°C (3 years) or 4°C (2 years).
  • Stable in solvent when stored at -80°C (1 year) or -20°C (6 months).
Catalog No. 50-002-12502 Supplier Medchemexpress LLC Supplier No. HY10129200MG
Veliparib (ABT-888) is a small-molecule inhibitor of poly(ADP-ribose) polymerases used in research to probe DNA repair pathways and for chemosensitization studies. It is supplied as a solid for analytical and preclinical research and has documented storage recommendations for powder and solutions.
  • Potent PARP1 and PARP2 inhibition (Ki 5.2 nM and 2.9 nM).
  • Molecular formula C13H16N4O and molecular weight 244.29 g/mol.
  • Identified by CAS number 912444-00-9.
  • Supplied as a powder with recommended storage: -20°C (3 years) or 4°C (2 years).
  • Stable in solvent when stored at -80°C (1 year) or -20°C (6 months).