Learn More
Medchemexpress LLC Veliparib dihydrochloride | 912445-05-7 | 100.0% | 317.21 g/mol | C13H18Cl2N4O | 200 MG
SDP
Catalog No. 5000211213
Encompass_Preferred
Veliparib dihydrochloride is the dihydrochloride salt of veliparib, a small-molecule inhibitor of poly(ADP-ribose) polymerases PARP1 and PARP2 used in biochemical and cell-based research to probe DNA damage response and repair pathways. Supplied as a high-purity solid, it is intended for mechanistic studies and preclinical research applications.
  • Potent inhibitor of PARP1 and PARP2 with Kis of 5.2 nM and 2.9 nM.
  • High reported purity suitable for research use (99.98%).
  • Well characterized molecular properties (MW 317.21 g/mol, formula C13H18Cl2N4O).
  • Suitable for biochemical and cell-based assays investigating DNA repair.
  • Available in milligram to gram quantities for diverse experimental needs.
Catalog No. 50-002-11213 Supplier Medchemexpress LLC Supplier No. HY10130200MG
Veliparib dihydrochloride is the dihydrochloride salt of veliparib, a small-molecule inhibitor of poly(ADP-ribose) polymerases PARP1 and PARP2 used in biochemical and cell-based research to probe DNA damage response and repair pathways. Supplied as a high-purity solid, it is intended for mechanistic studies and preclinical research applications.
  • Potent inhibitor of PARP1 and PARP2 with Kis of 5.2 nM and 2.9 nM.
  • High reported purity suitable for research use (99.98%).
  • Well characterized molecular properties (MW 317.21 g/mol, formula C13H18Cl2N4O).
  • Suitable for biochemical and cell-based assays investigating DNA repair.
  • Available in milligram to gram quantities for diverse experimental needs.