Learn More
Apexbio Technology LLC Veliparib dihydrochloride 912445-05-7 200mg
SDP
Catalog No. 5000064123
Encompass_Preferred
Veliparib dihydrochloride (CAS 912445-05-7) is a selective inhibitor of poly(ADP-ribose) polymerases PARP1 and PARP2 enzymes responsible for DNA repair processes frequently upregulated in various cancers Veliparib inhibits PARP1 and PARP2 with K(i) values of 5 2 and 2 9 nmol/L respectively In colon cancer cell lines (HCT-116 HT-29) veliparib enhances DNA damage and induces G2/M arrest when combined with SN38 or oxaliplatin In preclinical murine melanoma (B16F10) breast cancer (MX-1) and colon cancer (HCT-116) xenografts combination therapy with DNA-damaging agents shows improved antitumor effects
Catalog No. 50-000-64123 Supplier Apexbio Technology LLC Supplier No. A3958200
Veliparib dihydrochloride (CAS 912445-05-7) is a selective inhibitor of poly(ADP-ribose) polymerases PARP1 and PARP2 enzymes responsible for DNA repair processes frequently upregulated in various cancers Veliparib inhibits PARP1 and PARP2 with K(i) values of 5 2 and 2 9 nmol/L respectively In colon cancer cell lines (HCT-116 HT-29) veliparib enhances DNA damage and induces G2/M arrest when combined with SN38 or oxaliplatin In preclinical murine melanoma (B16F10) breast cancer (MX-1) and colon cancer (HCT-116) xenografts combination therapy with DNA-damaging agents shows improved antitumor effects