Learn More
MSC Vellumoid Gasket Sheet, Treated Cellulose Fiber, General Purpose, 250°F
Catalog No. 502835814
Encompass
Gasket sheet, made from treated cellulose fiber material, is impregnated with a protein glue and glycerin binder. This general-purpose gasket material offers reliable sealing in various applications, resisting oils, water coolants, and salt solutions. It is suitable for service temperatures up to 250°F and is commonly supplied in sheets of various dimensions to meet diverse industrial requirements.
  • Composed of treated cellulose fiber material.
  • Impregnated with protein glue and glycerin binder.
  • Designed for general purpose gasket applications.
  • Resistant to oils, water coolants, and salt solutions.
  • Operates effectively up to 250°F.
  • UL listed for verified compliance.
  • Provides good compressibility for effective sealing on uneven surfaces.
Catalog No. 50-283-5814 Supplier MSC Supplier No. 70909916
Gasket sheet, made from treated cellulose fiber material, is impregnated with a protein glue and glycerin binder. This general-purpose gasket material offers reliable sealing in various applications, resisting oils, water coolants, and salt solutions. It is suitable for service temperatures up to 250°F and is commonly supplied in sheets of various dimensions to meet diverse industrial requirements.
  • Composed of treated cellulose fiber material.
  • Impregnated with protein glue and glycerin binder.
  • Designed for general purpose gasket applications.
  • Resistant to oils, water coolants, and salt solutions.
  • Operates effectively up to 250°F.
  • UL listed for verified compliance.
  • Provides good compressibility for effective sealing on uneven surfaces.