Learn More
Medchemexpress LLC Velmupressin acetate | 1647120-04-4 | 99.5% | 1006.63 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1809A10MG
Encompass_Preferred
Velmupressin acetate is a potent, selective, and short-acting peptidic V2 receptor (V2R) agonist. It exhibits high affinity for both human (hV2R) and rat (rV2R) V2 receptors, with EC50s of 0.07 nM and 0.02 nM respectively. This product is intended for research purposes only.
  • Potent and selective V2 receptor agonist
  • Short-acting peptidic compound
  • High affinity for human and rat V2 receptors
  • For research purposes only
Catalog No. 50-004-22525