Learn More
Medchemexpress LLC Velnacrine maleate | 118909-22-1 | 98.0% | 330.34 | 25 MG
SDP
Catalog No. 5000366715
Encompass_Preferred
Velnacrine maleate (HP 029) is an orally active cholinesterase inhibitor used in the research of Alzheimer's disease. It is a solid appearing from light yellow to khaki color, and it is soluble in DMSO. This compound is provided with comprehensive data including a datasheet, COA, and SDS.
  • Orally active cholinesterase inhibitor
  • Can be used for Alzheimer's disease research
  • Comprehensive documentation including datasheet, COA, and SDS
  • Provided as a solid with color ranging from light yellow to khaki
  • Soluble in DMSO
Catalog No. 50-003-66715 Supplier Medchemexpress LLC Supplier No. 25MGHYW011246
Velnacrine maleate (HP 029) is an orally active cholinesterase inhibitor used in the research of Alzheimer's disease. It is a solid appearing from light yellow to khaki color, and it is soluble in DMSO. This compound is provided with comprehensive data including a datasheet, COA, and SDS.
  • Orally active cholinesterase inhibitor
  • Can be used for Alzheimer's disease research
  • Comprehensive documentation including datasheet, COA, and SDS
  • Provided as a solid with color ranging from light yellow to khaki
  • Soluble in DMSO