Learn More
Brady™ Velocity Alum Padlock 3 in. Stl Shkl KD 6/PK
Catalog No. 19221681 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-221-681 Yellow
19-221-457 Black
19-221-648 Blue
19-221-535 Brown
19-221-590 Green
19-221-411 Orange
19-221-607 Purple
19-221-626 Red
19-221-413 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-221-681 Supplier Brady™ Supplier No. SDALYLW76STKD6
Add to Cart
Add to Cart
Accelerate your program with durable products, available in the widest variety of colors, materials and styles, ideal for any size lockout program.
  • Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable and exclusively for safety
  • Color-matched keys and padlock bodies help workers find the right key for the right lock fast
  • Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities
  • Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection
  • Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access
  • Key codes engraved on the key and lock body to simplify record-keeping and traceability
  • Aluminum padlocks with steel shackles are corrosion-resistant and ideal for general lockout applications where added durability or tamper resistance are needed
Specifications
Color Yellow
Key Type Keyed Different
Material Nylon
Product Type Padlock