Description
- Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable and exclusively for safety
- Color-matched keys and padlock bodies help workers find the right key for the right lock fast
- Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection
- Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities
- Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access
- Key codes engraved on the key and lock body to simplify record-keeping and traceability
- Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking and ideal for electrical or general lockout applications
Specifications
Specifications
| Color | Yellow |
| Key Type | Keyed Alike |
| Material | Nylon |
| Product Type | Padlock |