Learn More
Brady™ Velocity Nylon Padlock 1.5 in. Pla Shkl KD 6/Pk.
Catalog No. 19221682 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Brown
Purple
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-221-682 Purple
19-221-610 Black
19-221-579 Brown
19-221-615 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-221-682 Supplier Brady™ Supplier No. SDPLPRP38PLKD6
Add to Cart
Add to Cart
Accelerate your program with durable products available in the widest variety of colors, materials and styles, ideal for any size lockout program.
  • Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable and exclusively for safety
  • Color-matched keys and padlock bodies help workers find the right key for the right lock fast
  • Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection
  • Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities
  • Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access
  • Key codes engraved on the key and lock body to simplify record-keeping and traceability
  • Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking and ideal for electrical or general lockout applications
Specifications
Color Purple
Key Type Keyed Different
Material Nylon
Product Type Padlock