Learn More
Brady™ Velocity Nylon Padlock 1.5 in. Stl Shkl KA 3/Pk.
Catalog No. 19221661 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-221-661 Black
19-221-611 Blue
19-221-580 Brown
19-221-594 Green
19-221-653 Orange
19-221-380 Purple
19-221-609 Red
19-221-415 White
19-221-596 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-221-661 Supplier Brady™ Supplier No. SDPLBLK38STKA3
Add to Cart
Add to Cart
Accelerate your program with durable products available in the widest variety of colors, materials and styles, ideal for any size lockout program.
  • Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable and exclusively for safety
  • Color-matched keys and padlock bodies help workers find the right key for the right lock fast
  • Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities
  • Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection
  • Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access
  • Key codes engraved on the key and lock body to simplify record-keeping and traceability
  • Nylon padlocks with steel shackles are lightweight, durable and ideal for general lockout applications
Specifications
Color Black
Key Type Keyed Alike
Material Nylon
Product Type Padlock