Learn More
Brady™ Velocity Nylon Padlock 1 in. Pla Shkl KD 1/Ea.
Catalog No. 19221695 Shop All Brady Corporation Products
Change view
Click to view available options
Color:
Black
Blue
Brown
Green
Orange
Purple
Red
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-221-695 Red
19-221-545 Black
19-221-506 Blue
19-221-679 Brown
19-221-426 Green
19-221-642 Orange
19-221-536 Purple
19-221-693 White
19-221-434 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-221-695 Supplier Brady™ Supplier No. CPPLRED25PLKD
Add to Cart
Add to Cart
Compact design ideal for lockout applications where clearance or accessibility is limited.
  • Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable and exclusively for safety
  • Color-matched keys and padlock bodies help workers find the right key for the right lock fast
  • Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities
  • Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection
  • Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access
  • Key codes engraved on the key and lock body to simplify record-keeping and traceability
  • Nylon padlocks with plastic shackles are lightweight, non-conductive, non-sparking and ideal for electrical or general lockout applications
Specifications
Color Red
Key Type Keyed Different
Material Nylon
Product Type Padlock