Learn More
Cole-Parmer Cole-Parmer Airflow Meter Vaneometer
Supplier:  Cole-Parmer 7890012
Encompass_Preferred
Continuously measure velosity of air flow into hood to determine when to change filters. Measure low air velocities from 25 to 400 FPM. Easy to use, even by untrained personnel.
Catalog No. NC1906078