Learn More
Sentry Air Systems Velometer
SDP
Catalog No. NC0311834
Encompass
The Velometer measures the airflow of the unit in feet/minute. This accessory helps the operator know the airflow speed of the current fan setting which is helpful for units with a variable speed control.
Catalog No. NC0311834 Supplier Sentry Air Systems Supplier No. SS000VEL
The Velometer measures the airflow of the unit in feet/minute. This accessory helps the operator know the airflow speed of the current fan setting which is helpful for units with a variable speed control.