Learn More
Cole-Parmer Velp F203A0179 Multiposition Digital Stirrer, 6-position, 120-240V
Supplier:  Cole-Parmer 8657957
Encompass
Excellent speed control and synchronized stirring ensure that the same speed is achieved on each position even at low speeds. The low-profile, lightweight design makes these stirrers easy to move while saving valuable bench space. Stainless steel top plate construction is easy to clean and provides high chemical resistance. Featuring adjustable time controlled reverse operation, timer, and a bright digital display for accurate speed monitoring, these stirrers will provide years of worry-free operation.
Catalog No. NC1141708