Learn More
Velp™ OXISoft™ Software
Description
- This software manages all analysis steps and readily provides valuable information to scientists conducting experiments
- Enables scientists to perform repeatability tests, freshness tests, formula comparison, packaging comparison, IP during ageing, and estimated shelf life
- Up to four tests can be managed by the same software for maximum productivity
Specifications
Specifications
| For Use With (Equipment) | OHS Controllers |
| Type | Software |
| Voltage | 115/230 VAC |