Learn More
Selleck Chemical LLC Velpatasvir 25mg 1377049-84-7 GS-5816
SDP
Supplier:  Selleck Chemical LLC S372425MG
Encompass_Preferred
Velpatasvir (GS-5816) is a second-generation NS5A inhibitor that inhibits hepatitis C viral replication through acting on the crucial "membranous web" that facilitates RNA replication. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8317