Learn More
Cayman Chemical VelpatsvIr 100mg
SDP
Catalog No. 502981429
Encompass_Preferred
An NS5A inhibitor; inhibits replication of HCV genotypes 1-6 (EC50s = 6-130 pM in replicon assays); inhibits SARS-CoV-2 exonuclease activity at 100 µM; enhances remdesivir-induced inhibition of SARS-CoV-2 replication in infected Calu-3 cells at 10 µM
Catalog No. 50-298-1429 Supplier Cayman Chemical Supplier No. 21280100 MG
An NS5A inhibitor; inhibits replication of HCV genotypes 1-6 (EC50s = 6-130 pM in replicon assays); inhibits SARS-CoV-2 exonuclease activity at 100 µM; enhances remdesivir-induced inhibition of SARS-CoV-2 replication in infected Calu-3 cells at 10 µM