Learn More
Medchemexpress LLC Velsecorat | 1196509-60-0 | 99.8% | 606.62 | 100 MG
SDP
Catalog No. 5000345554
Encompass_Preferred
Velsecorat (AZD7594) is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM. It shows no effect on progesterone receptor, mineralocorticoid receptor, androgen receptor, ERα or ERβ (IC50, > 10 μM). This compound has anti-inflammatory activity and efficiently inhibits rat lung edema.
  • Potent selective nonsteroidal glucocorticoid receptor modulator
  • IC50 of 0.9 nM (Glucocorticoid receptor)
  • Shows no effect on progesterone receptor, mineralocorticoid receptor, androgen receptor, ERα or ERβ (IC50, > 10 μM)
  • Anti-inflammatory activity
  • Efficiently inhibits rat lung edema
This item is not returnable. View return policy
Catalog No. 50-003-45554 Supplier Medchemexpress LLC Supplier No. HY11145310MG
Velsecorat (AZD7594) is a potent selective nonsteroidal glucocorticoid receptor modulator, with an IC50 of 0.9 nM. It shows no effect on progesterone receptor, mineralocorticoid receptor, androgen receptor, ERα or ERβ (IC50, > 10 μM). This compound has anti-inflammatory activity and efficiently inhibits rat lung edema.
  • Potent selective nonsteroidal glucocorticoid receptor modulator
  • IC50 of 0.9 nM (Glucocorticoid receptor)
  • Shows no effect on progesterone receptor, mineralocorticoid receptor, androgen receptor, ERα or ERβ (IC50, > 10 μM)
  • Anti-inflammatory activity
  • Efficiently inhibits rat lung edema