Learn More
VAI DECON-HAND™ Hand Sanitizer
Catalog No. 19546838 Shop All VAI Products
Change view
Click to view available options
Volume (English):
16 oz.
32 oz.
Sterility:
Non-sterile
Sterile
Container Type:
Bottle
Bottle for ASEPTI-CLEANSE Dispenser
Bottle with Attachable Pump
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Sterility Container Type
19-546-838 16 oz. Non-sterile Bottle with Attachable Pump
19-546-839 16 oz. Sterile Bottle with Attachable Pump
19-546-840 32 oz. Sterile Bottle
19-546-842 16 oz. Non-sterile -
19-546-775 32 oz. Non-sterile Bottle for ASEPTI-CLEANSE Dispenser
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-546-838 Supplier VAI Supplier No. DH04
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Lot-specific sterile documentation
Gelled alcohol DECON-HAND™ Hand Sanitizer decreases bacteria and fungi on skin.
  • An instant hand sanitizer before glove donning
  • Apply to hands and allow to dry without wiping
  • Provides effective sanitization against bacteria and fungi according to USP 25 criteria
  • Validated for sterility and shelf life
Compliance
Manufactured in accordance with 21 CFR Part 211 Good Manufacturing Practices for DRUGS and the Tentative Final Monograph for Topical Antimicrobial Drug Products for Over-the-Counter human use.
Specifications
Product Type Hand Sanitizer
Sterility Non-sterile
Container Type Bottle with Attachable Pump
For Use With (Application) For use as an instant hand sterilizer before glove donning
Volume (English) 16 oz.
Volume (Metric) 473 mL