Learn More
Medchemexpress LLC HY-12057 50mg Medchemexpress, Vemurafenib CAS:918504-65-1 Purity:>98%
SDP
Catalog No. 502028849
Encompass_Preferred
Medchemexpress, HY-12057 50mg Vemurafenib CAS:918504-65-1 Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively[4]. Vemurafenib induces cell autophagy[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8849 Supplier Medchemexpress LLC Supplier No. HY1205750MG
Medchemexpress, HY-12057 50mg Vemurafenib CAS:918504-65-1 Vemurafenib (PLX4032) is a first-in-class, selective, potent inhibitor of B-RAF kinase, with IC50s of 31 and 48 nM for RAFV600E and c-RAF-1, respectively[4]. Vemurafenib induces cell autophagy[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.