Learn More
Apexbio Technology LLC Vemurafenib (PLX4032, RG7204), 500mg. Cas: 918504-65-1 MFCD: MFCD18074504. BRAF kinase inhibitor
SDP
Supplier:  Apexbio Technology LLC A3004500
Encompass_Preferred
Vemurafenib is an inhibitor of BRAF kinase. It inhibits BRAFV600E and also has inhibitory activity in vitro against several other kinds of kinases, including CRAF, ARAF and wild-type BRAF Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4774