Learn More
Selleck Chemical LLC Vemurafenib (PLX4032) 50mg 918504-65-1 RG7204
SDP
Catalog No. 508461
Encompass_Preferred
Vemurafenib (PLX4032, RG7204) is a novel and potent inhibitor of B-RafV600E with IC50 of 31 nM in cell-free assay. 10-fold selective for B-RafV600E over wild-type B-Raf in enzymatic assays and the cellular selectivity can exceed 100-fold. Vemurafenib (PLX4032, RG7204) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-846-1 Supplier Selleck Chemical LLC Supplier No. S126750MG
Add to Cart
Add to Cart
Vemurafenib (PLX4032, RG7204) is a novel and potent inhibitor of B-RafV600E with IC50 of 31 nM in cell-free assay. 10-fold selective for B-RafV600E over wild-type B-Raf in enzymatic assays and the cellular selectivity can exceed 100-fold. Vemurafenib (PLX4032, RG7204) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.