Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC VENAIR .25ID X .375OD 50FTROLL
Catalog No. NC2049484
Encompass_Preferred
Venair .25 in ID x .375 in OD, platinum cured silicone tubing, 60shA, 50 foot roll
Catalog No. NC2049484 Supplier ATLANTIC BIOPHARM SOLUTIONS LLC Supplier No. ABS920085200609
Venair .25 in ID x .375 in OD, platinum cured silicone tubing, 60shA, 50 foot roll