Learn More
MSC MGK Cockroach Gel Bait, 0.50% Clothianidin, 0.50% Pyriproxyfen, 4 x 30 g Tubes
Catalog No. 502836757
Encompass
This professional-grade gel bait attracts and eliminates both bait-averse and non-averse cockroaches. It features a palatable bait matrix stable in high temperatures, ideal for diverse environments including commercial, industrial, residential, food processing, and stored food areas. Contains 0.50% Clothianidin and 0.50% Pyriproxyfen, supplied in four 30 gram tubes.
  • Palatable bait matrix
  • Attracts and kills bait averse and non-averse cockroaches
  • Fits standard bait guns
  • Consistent performance across dietary conditions
  • Clean, odorless bait; stable in high heat
  • For commercial, industrial, residential, food, and non-food areas
  • Free of common allergens (milk, eggs, fish, shellfish, tree nuts, wheat)
  • Easy to handle and apply
Catalog No. 50-283-6757 Supplier MSC Supplier No. 74320284
Add to Cart
Add to Cart
This professional-grade gel bait attracts and eliminates both bait-averse and non-averse cockroaches. It features a palatable bait matrix stable in high temperatures, ideal for diverse environments including commercial, industrial, residential, food processing, and stored food areas. Contains 0.50% Clothianidin and 0.50% Pyriproxyfen, supplied in four 30 gram tubes.
  • Palatable bait matrix
  • Attracts and kills bait averse and non-averse cockroaches
  • Fits standard bait guns
  • Consistent performance across dietary conditions
  • Clean, odorless bait; stable in high heat
  • For commercial, industrial, residential, food, and non-food areas
  • Free of common allergens (milk, eggs, fish, shellfish, tree nuts, wheat)
  • Easy to handle and apply