Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Vendsorb Chemical Absorbent Rolls

Catalog No. 17111168 Shop All Brady Corporation Products
Change view
Click to view available options
Dimensions (L x W):
12 x 60 in.
19.05 x 127 cm (7.5 x 50 in.)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
17-111-168 12 x 60 in.
17-111-169 19.05 x 127 cm (7.5 x 50 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-111-168 Supplier Brady™ Supplier No. VSH12P

Designed to absorb hazardous chemicals and acids

  • Conveniently sized
  • Chemically inert making it ideal for aggressive fluids
  • Perforated and dimpled
  • Available in two sizes: 12 x 60 in. and 7.5 x 50 in.

Specifications

Type Chemical Absorbent Roll
Packaging Quantity 20/Cs.
Dimensions (L x W) 12 x 60 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.