Learn More
Medchemexpress LLC HY-15531 100mg , Venetoclax CAS:1257044-40-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15531100MG
Encompass_Preferred
HY-15531 100mg Venetoclax CAS: 1257044-40-8 Venetoclax (ABT-199; GDC-0199) is a highly potent, selective and orally bioavailable Bcl-2 inhibitor with a Ki of less than 0.01 nM. Venetoclax induces autophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8954