Learn More
Enzo Life Sciences Venetoclax (1g). CAS: 1257044-40-8
SDP
Catalog No. 502013692
Encompass_Preferred
Venetoclax is a Bcl-2-selective inhibitor with Ki of <0.01 nM, >4800-fold more selective versus Bcl-xL and Bcl-w, and no activity to Mcl-1 Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-3692 Supplier Enzo Life Sciences Supplier No. ENZCHM2411000
Venetoclax is a Bcl-2-selective inhibitor with Ki of <0.01 nM, >4800-fold more selective versus Bcl-xL and Bcl-w, and no activity to Mcl-1 Purity: ≥99% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.