Learn More
Medchemexpress LLC Venetoclax-d8 (ABT-199-d8) | 1257051-06-1 | 99.8% | 876.49 g/mol | C45H42D8ClN7O7S | 10 MG
SDP
Catalog No. 5000286839
Encompass_Preferred
Venetoclax-d8 is a deuterium-labeled isotopic standard of the Bcl-2 inhibitor venetoclax, supplied as a pre-weighed reference material for research and analytical workflows including mass spectrometry, method development, and pharmacokinetic studies.
  • Deuterium-labeled (d8) isotopic standard for internal standardization.
  • High isotopic and chemical purity for accurate quantification.
  • Pre-weighed, solid material for reproducible dosing and preparation.
  • Suitable for mass spectrometry and pharmacokinetic analyses.
  • Available in multiple small package sizes for analytical use.
Catalog No. 50-002-86839 Supplier Medchemexpress LLC Supplier No. HY15531S10MG
Add to Cart
Add to Cart
Venetoclax-d8 is a deuterium-labeled isotopic standard of the Bcl-2 inhibitor venetoclax, supplied as a pre-weighed reference material for research and analytical workflows including mass spectrometry, method development, and pharmacokinetic studies.
  • Deuterium-labeled (d8) isotopic standard for internal standardization.
  • High isotopic and chemical purity for accurate quantification.
  • Pre-weighed, solid material for reproducible dosing and preparation.
  • Suitable for mass spectrometry and pharmacokinetic analyses.
  • Available in multiple small package sizes for analytical use.