Learn More
Apexbio Technology LLC Venlafaxine 99300-78-4 100mg
SDP
Catalog No. 502240486
Encompass_Preferred
Venlafaxine (CAS 99300-78-4) is a small molecule characterized as a dual inhibitor of serotonin and noradrenaline reuptake transporters It exhibits approximately 30-fold greater affinity for the serotonin transporter (SERT K sub i /sub 82 nM) relative to the noradrenaline transporter (NET K sub i /sub 2480 nM) In preclinical models venlafaxine has been shown to enhance swimming and climbing behaviors in the rat forced-swim test indicative of antidepressant-like activity This compound is utilized in research settings to investigate the pharmacological modulation of monoaminergic neurotransmission and the neurobiological mechanisms underlying antidepressant responses
This item is not returnable. View return policy
Catalog No. 50-224-0486 Supplier Apexbio Technology LLC Supplier No. A5355100
Venlafaxine (CAS 99300-78-4) is a small molecule characterized as a dual inhibitor of serotonin and noradrenaline reuptake transporters It exhibits approximately 30-fold greater affinity for the serotonin transporter (SERT K sub i /sub 82 nM) relative to the noradrenaline transporter (NET K sub i /sub 2480 nM) In preclinical models venlafaxine has been shown to enhance swimming and climbing behaviors in the rat forced-swim test indicative of antidepressant-like activity This compound is utilized in research settings to investigate the pharmacological modulation of monoaminergic neurotransmission and the neurobiological mechanisms underlying antidepressant responses