Learn More
Medchemexpress LLC Venlafaxine-d10 hydrochloride | 1216539-56-8 | >98% | 2.5 MG
SDP
Catalog No. 5000406624
Encompass_Preferred
Venlafaxine-d10 hydrochloride is a deuterium labeled version of Venlafaxine hydrochloride, an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor, and an antidepressant. Intended for research use only, its deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Venlafaxine hydrochloride
  • Potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antidepressant properties
Catalog No. 50-004-06624 Supplier Medchemexpress LLC Supplier No. HYB0196AS2.5MG
Venlafaxine-d10 hydrochloride is a deuterium labeled version of Venlafaxine hydrochloride, an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor, and an antidepressant. Intended for research use only, its deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled Venlafaxine hydrochloride
  • Potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antidepressant properties