Learn More
Medchemexpress LLC Venlafaxine-d6 hydrochloride | 1062606-12-5 | 98.2% | 319.90 | C17H22D6ClNO2 | 10 MG
SDP
Catalog No. 5000298289
Encompass_Preferred
Venlafaxine-d6 hydrochloride is the deuterium-labeled hydrochloride salt of venlafaxine, provided as a high-purity analytical standard for bioanalytical and mass spectrometry applications. It is intended for use as an internal standard in quantitative assays to improve accuracy and precision. The substance has CAS 1062606-12-5 and molecular weight 319.90.
  • Deuterium-labeled internal standard for LC-MS/MS quantitation
  • High purity (98.2%) for reliable analytical performance
  • Hydrochloride salt form, stable under typical storage conditions
  • Suitable for pharmacokinetic and toxicology studies
  • Supplied in verified small quantities for reference use
Catalog No. 50-002-98289 Supplier Medchemexpress LLC Supplier No. HYB0196AS110MG
Venlafaxine-d6 hydrochloride is the deuterium-labeled hydrochloride salt of venlafaxine, provided as a high-purity analytical standard for bioanalytical and mass spectrometry applications. It is intended for use as an internal standard in quantitative assays to improve accuracy and precision. The substance has CAS 1062606-12-5 and molecular weight 319.90.
  • Deuterium-labeled internal standard for LC-MS/MS quantitation
  • High purity (98.2%) for reliable analytical performance
  • Hydrochloride salt form, stable under typical storage conditions
  • Suitable for pharmacokinetic and toxicology studies
  • Supplied in verified small quantities for reference use