Learn More
TARGETMOL CHEMICALS INC VENLAFAXINE HYDROCHLORID 200MG
SDP
Catalog No. 502581784
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Venlafaxine hydrochloride (Wy 45030 hydrochloride) is a cyclohexanol and phenylethylamine derivative that functions as a SEROTONIN AND NORADRENALINE REUPTAKE INHIBITOR (SNRI) and is used as an ANTIDEPRESSIVE AGENT. purity: 99%
Catalog No. 50-258-1784 Supplier TARGETMOL CHEMICALS INC Supplier No. T0472200MG
Also available in 5 mg 10 mg 25 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Venlafaxine hydrochloride (Wy 45030 hydrochloride) is a cyclohexanol and phenylethylamine derivative that functions as a SEROTONIN AND NORADRENALINE REUPTAKE INHIBITOR (SNRI) and is used as an ANTIDEPRESSIVE AGENT. purity: 99%