Learn More
Medchemexpress LLC Venlafaxine hydrochloride | 99300-78-4 | MFCD03658865 | 99.8% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0196A50MG
Encompass_Preferred
Venlafaxine hydrochloride is an orally active, potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor and an antidepressant, primarily used for research purposes. It targets the Serotonin Transporter in neuronal signaling pathways.
  • Potent serotonin (5-HT)/norepinephrine (NE) reuptake dual inhibitor.
  • Antidepressant properties.
  • Active orally.
  • Used for research only.
Catalog No. 50-004-05800